VWR® AVS TITRINORM, Germanium Standard Solution, 1000 mg/L Ge in 5% Nitric Acid with hydrofluoric Acid (max. 1%) AVS TITRINORM Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86680.180EA 36 EUR
86680.180 86680.260
VWR® AVS TITRINORM, Germanium Standard Solution, 1000 mg/L Ge in 5% Nitric Acid with hydrofluoric Acid (max. 1%) AVS TITRINORM Standard for AAS
Standaarden Kationische standaarden Germanium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%