VWR®, Niobium standard solution, 1000 mg/L Nb in 5% nitric acid with hydrofluoric acid (max. 1%) AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86694.180EA 29.8 EUR
86694.180 86694.260
VWR®, Niobium standard solution, 1000 mg/L Nb in 5% nitric acid with hydrofluoric acid (max. 1%) AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Niobium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%