Ethyl-3-hydroxybutyrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate

H0230-25G H0230-500G
TCIAH0230-25GEA 18.7 EUR
TCIAH0230-25G TCIAH0230-500G
Ethyl-3-hydroxybutyrate ≥98.0% (by GC)
Ethyl-3-hydroxybutyrate
Formule: C₆H₁₂O₃
MW: 132.16 g/mol
Kookpunt: 180°C
Vlampunt: 64°C
MDL Nummer: MFCD00004545
CAS nummer: 5405-41-4
EINECS: 226-456-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70361ea9578ed11b244549ec9980c2c59d68565f&vid=70361ea9578ed11b244549ec9980c2c59d68565f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR