3-Ethyl-3-oxetanemethanol

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (3-Ethyloxetan-3-yl)methanol, 2-Hydroxymethyl-2-ethyl-1,3-epoxypropane, 3-Hydroxymethyl-3-ethyloxetane

E0546-25G E0546-500G E0546-100G
TCIAE0546-25GEA 31.9 EUR
TCIAE0546-25G TCIAE0546-500G TCIAE0546-100G
3-Ethyl-3-oxetanemethanol
3-Ethyl-3-oxetanemethanol
Formule: C₆H₁₂O₂
MW: 116.16 g/mol
Kookpunt: 110°C (literatuur)
Vlampunt: 112°C (literatuur)
MDL Nummer: MFCD00190143
CAS nummer: 3047-32-3
EINECS: 221-254-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45de692da41ef548f48425ee36d381edf09823a1&vid=45de692da41ef548f48425ee36d381edf09823a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR