2-(Allylthio)nicotinic acid
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB06373.1
BTB06373.10
BTB06373.50
MAYBBTB06373.1EA
0
EUR
MAYBBTB06373.1
MAYBBTB06373.10
MAYBBTB06373.50
2-(Allylthio)nicotinic acid
2-(Allylthio)nicotinic acid
Formule:
C₉H₉NO₂S MW: 195.24 g/mol |
MDL Nummer:
MFCD00051681 CAS nummer: 175135-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...