Chloromethanesulphonyl chloride ≥95.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C0924-25G
C0924-5G
TCIAC0924-25GEA
206
EUR
TCIAC0924-25G
TCIAC0924-5G
Chloromethanesulphonyl chloride ≥95.0% (by titrimetric analysis)
Chloromethanesulphonyl chloride
Formule:
CH₂Cl₂O₂S Kookpunt: 175°C |
MDL Nummer:
MFCD00014727 CAS nummer: 3518-65-8 EINECS: 222-526-1 UN: 3265 ADR: 8,II |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (Argentmetric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...