Ethyl-4-piperidinecarboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0294-25ML I0294-500ML I0294-100ML
TCIAI0294-25MLEA 27.5 EUR
TCIAI0294-25ML TCIAI0294-500ML TCIAI0294-100ML
Ethyl-4-piperidinecarboxylate ≥98.0% (by GC)
Ethyl-4-piperidinecarboxylate
Formule: C₈H₁₅NO₂
Kookpunt: 105°C
Vlampunt: 80°C
MDL Nummer: MFCD00006003
CAS nummer: 1126-09-6
EINECS: 214-416-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=909d577793214417669290e45eabf84695437b8f&vid=909d577793214417669290e45eabf84695437b8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR