6-Bromochromone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3335-25G B3335-5G
TCIAB3335-25GEA 242 EUR
TCIAB3335-25G TCIAB3335-5G
6-Bromochromone ≥98.0% (by GC)
6-Bromochromone
Formule: C₉H₅BrO₂
MDL Nummer: MFCD00239369
CAS nummer: 51483-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f74fad0117c89f81ae7b83dd86a4bf04796936d&vid=8f74fad0117c89f81ae7b83dd86a4bf04796936d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR