N-Oleoyl taurine
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10005609-1 10005609-10 10005609-5 10005609-500
CAYM10005609-1EA 124 EUR
CAYM10005609-1 CAYM10005609-10 CAYM10005609-5 CAYM10005609-500
N-Oleoyl taurine
N-Oleoyl taurine
Formule: C₂₀H₃₉NO₄S
CAS nummer: 52514-04-2
EINECS: 257-985-7