Ethyl-2-hydroxyisobutyrate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
H0255-25G H0255-250G
TCIAH0255-25GEA 66 EUR
TCIAH0255-25G TCIAH0255-250G
Ethyl-2-hydroxyisobutyrate ≥98.0% (by GC)
Ethyl-2-hydroxyisobutyrate
Formule: C₆H₁₂O₃
Kookpunt: 150°C
Vlampunt: 41°C
MDL Nummer: MFCD00004458
CAS nummer: 80-55-7
EINECS: 201-290-3
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95606df4ff5292c1149a0b50462f3dd710a25fa2&vid=95606df4ff5292c1149a0b50462f3dd710a25fa2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %