2-Methoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M2159-1G
M2159-5G
TCIAM2159-1GEA
27.5
EUR
TCIAM2159-1G
TCIAM2159-5G
2-Methoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
2-Methoxy-3-pyridineboronic acid
Formule:
C₆H₈BNO₃ Smeltpunt: 145°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD03411572 CAS nummer: 163105-90-6 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...