Bis(tri-tert-butylphosphine)palladium(0)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)
Packaged under inert gas.
44845.03
44845.01
44845.03EA
278
EUR
44845.03
44845.01
Bis(tri-tert-butylphosphine)palladium(0)
Bis(tri-tert-butylphosphine)palladium(0)
CAS No.: 53199-31-8
Molecular Formula: C24H54P2Pd
Formula Weight: 511.06
Storage Temperature: -30°C to -10°C
Physical Form: Powder, packaged under inert atmosphere
Sensitivity: Light, Air and Moisture Sensitive
Appearance: White to off-white
Melting Point: 258-272°
MDL No.: MFCD03094580
Coupling reactions, Heck couplings
Formule:
C₂₄H₅₄P₂Pd MW: 511.05 g/mol Smeltpunt: 258…272 °C Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD03094580 CAS nummer: 53199-31-8 |