EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4015690.0100 4015690.0500
BACE4015690.0100EA 0 EUR
BACE4015690.0100 BACE4015690.0500
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
Formule: C₁₄H₁₇NO₃
MDL Nummer: MFCD00006703
CAS nummer: 16357-59-8
EINECS: 240-418-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR