9-Bromo-9-phenylfluorene 96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 9-Bromo-9-phenyl-9H-fluorene

L14225.03 L14225.06
L14225.03EA 36.8 EUR
L14225.03 L14225.06
9-Bromo-9-phenylfluorene 96%
9-Bromo-9-phenylfluorene

Purity: 96% CAS No.: 55135-66-5 Molecular Formula: C19H13Br Formula Weight: 321.23 Melting Point: 94-99° Beilstein Reference: 2561445 UN No.: 3261 MDL No.: MFCD00075522

Formule: C₁₉H₁₃Br
MW: 321.23 g/mol
Smeltpunt: 94…99 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075522
CAS nummer: 55135-66-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR