3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB00576DA SB00576EA SB00576.50
MAYBSB00576DAEA 0 EUR
MAYBSB00576DA MAYBSB00576EA MAYBSB00576.50
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Formule: C₁₁H₆Cl₃NO₂
MDL Nummer: MFCD00052556
CAS nummer: 4462-55-9
EINECS: 224-723-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR