3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB00578.1 SB00578.10 SB00578.50
MAYBSB00578.1EA 0 EUR
MAYBSB00578.1 MAYBSB00578.10 MAYBSB00578.50
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride
Formule: C₁₁H₆Cl₂FNO₂
MW: 274.08 g/mol
MDL Nummer: MFCD00055650
CAS nummer: 69399-79-7
EINECS: 273-987-0
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR