3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbothioamide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RF00747.1 RF00747.10
MAYBRF00747.1EA 0 EUR
MAYBRF00747.1 MAYBRF00747.10
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbothioamide
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbothioamide
Formule: C₁₁H₈ClFN₂S
MDL Nummer: MFCD00052560
CAS nummer: 175204-42-9