4-(Trifluoromethylthio)phenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01940DA SB01940EA
MAYBSB01940DAEA 0 EUR
MAYBSB01940DA MAYBSB01940EA
4-(Trifluoromethylthio)phenol
4-(Trifluoromethylthio)phenol
Formule: C₇H₅F₃OS
MDL Nummer: MFCD00036035
CAS nummer: 461-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR