VWR® AVS TITRINORM, Zirconium Standard Solution, 1000 mg/L Zr in 5% Hydrochloric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86726.180EA
71.9
EUR
86726.180
86726.260
VWR® AVS TITRINORM, Zirconium Standard Solution, 1000 mg/L Zr in 5% Hydrochloric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
Standaarden
Kationische standaarden
Zirkonium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%