2-[4-(Bromomethyl)phenyl]benzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Methyl 2-[4-(Bromomethyl)phenyl]benzoate

M1806-25G M1806-5G
TCIAM1806-25GEA 69.3 EUR
TCIAM1806-25G TCIAM1806-5G
2-[4-(Bromomethyl)phenyl]benzoate ≥98.0% (by GC)
2-[4-(Bromomethyl)phenyl]benzoate
Formule: C₁₅H₁₃BrO₂
MDL Nummer: MFCD06200816
CAS nummer: 114772-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c59edb39fee4f9ed67f6f7b7c6a27ec8ee8132&vid=95c59edb39fee4f9ed67f6f7b7c6a27ec8ee8132&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR