Chromone-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

O0290-25G O0290-5G
TCIAO0290-25GEA 568 EUR
TCIAO0290-25G TCIAO0290-5G
Chromone-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Chromone-2-carboxylic acid
Formule: C₁₀H₆O₄
MDL Nummer: MFCD00006838
CAS nummer: 4940-39-0
EINECS: 225-583-0
Merck Index: 14,02237
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cad1685afcf07e276a399eab9e30652e1daec746&vid=cad1685afcf07e276a399eab9e30652e1daec746&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR