VWR®, Tungsten standard solution, 1000 mg/L W in nitric acid with hydrofluoric acid (each max. 1%) AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86722.260EA 58.3 EUR
86722.260 86722.180
VWR®, Tungsten standard solution, 1000 mg/L W in nitric acid with hydrofluoric acid (each max. 1%) AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Wolfraam standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%