VWR®, Rhenium standard solution, 1,000 mg/l Re in dil. nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86703.180EA
79.8
EUR
86703.180
86703.260
VWR®, Rhenium standard solution, 1,000 mg/l Re in dil. nitric acid AVS TITRINORM standard for AAS
Standaarden
Kationische standaarden
Renium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%