Ammonium 1-pyrrolidinecarbodithioate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
B23731.14 B23731.22
B23731.14EA 43.1 EUR
B23731.14 B23731.22
Ammonium 1-pyrrolidinecarbodithioate ≥98%
Ammonium 1-pyrrolidinecarbodithioate
Purity: 98% CAS No.: 5108-96-3 Molecular Formula: C5H12N2S2 Formula Weight: 164.29 Melting Point: 150-155° Beilstein Reference: 3730472
Formule: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Smeltpunt: 150…155 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012720
CAS nummer: 5108-96-3
EINECS: 225-834-4