Pipette tips for sample preparation, ZipTip®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ZTC18S008 ZTC18S960 ZTC18M096 ZTC18S096 ZTC04S096 ZTSCXS096 ZTC04S008 ZTC04S960 ZTC18M008 ZTC18M960 ZTSCXS008
MILFZTC18S008EA 59.8 EUR
MILFZTC18S008 MILFZTC18S960 MILFZTC18M096 MILFZTC18S096 MILFZTC04S096 MILFZTSCXS096 MILFZTC04S008 MILFZTC04S960 MILFZTC18M008 MILFZTC18M960 MILFZTSCXS008
Pipette tips for sample preparation, ZipTip®
Pipettips
ZipTip is a 10 µl pipette tip with a 0,6 or 0,2 µl bed of chromatography media fixed at its end with no dead volume. It is ideal for concentrating and purifying samples for sensitive downstream analyses.
  • Single-step desalting, concentration, and purification
  • Fractionate complex samples for more meaningful data
  • Ideal for peptides, proteins, nucleic acids, and more
  • No dead volume for maximum recovery
  • Eliminates time-consuming chromatography
The ZipTip pipette tip provides a reproducible, high-recovery method for concentrating and purifying femtomoles to picomoles of peptides, proteins and oligonucleotides for improved analytical data.
Available with three different ZipTip resins:
C18 Resin (0,6 µl or 0,2 µl bed volumes): For oligonucleotides or small (<50 kDa) proteins/peptides in aqueous solution
C4 Resin: For proteins or peptides (3 kDa to 100 kDa) in aqueous solution
Strong cationic resin (SCX): For peptides in solution with organic solvents or detergents