Dropping funnels, cylindrical, with pressure balance
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
443533 443540 443547 443554
201-0621EA 190 EUR
201-0621 201-0622 201-0623 201-0624
Dropping funnels, cylindrical, with pressure balance
Trechters Druppeltrechters
PYREX®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For use when liquids have to be added to systems under vacuum. Double white graduated scale showing volume of liquid contained/delivered. Standard ground joint cone and socket.