Spherical ground joints, Rotulex®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RM19 RM29 RF13 RF19 RF29 RF41 RM13 RO13 RO19 RO29 RO35 RO41 RM41
BIBBRM19EA 119 EUR
BIBBRM19 BIBBRM29 201-8212 201-8213 201-8214 BIBBRF41 BIBBRM13 BIBBRO13 BIBBRO19 BIBBRO29 BIBBRO35 BIBBRO41 BIBBRM41
Spherical ground joints, Rotulex®
Adapters Adapters met kogelgewrichten
Made of borosilicate glass.
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200 °C
  • System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
  • Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
Bestelinformatie: Component parts should be ordered separately .