Screwthread joints
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SQ13UB SQ18UB SQ24UB SQ28UB
BIBBSQ13UBEA 72.2 EUR
BIBBSQ13UB BIBBSQ18UB BIBBSQ24UB BIBBSQ28UB
Screwthread joints
Adapters Adapters, Buizen met schroefdraad
Pyrex®, borosilicate glass.
  • Resistance to thermal shock and chemical corrosion
  • Designed to use with the range of plastic screwcaps, silicone rubber rings and PTFE washers
  • Unbadged
It is used by glassblowers in the construction of laboratory glassware.