Torion stopcocks, SLV®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
722-53 722-52 722-41 722-42 722-51 722-43
BIBB722-53EA 1330 EUR
BIBB722-53 BIBB722-52 BIBB722-41 BIBB722-42 BIBB722-51 BIBB722-43
Torion stopcocks, SLV®
Afsluitkranen
Stopcocks are made of borosilicate glass.
  • Vacuum capability to 10⁻² Torr
  • Sidearms are 70 mm in length
  • Excellent chemical and temperature resistance
In right angle pattern PTFE key is screwed into the threaded Torion connector, to open and close the stopcock.
Leveringsinformatie: Supplied with fittings, sealing rings and PTFE key.