Wash bottles, wide neck, DIY, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WGW001PML WGW002PML
215-3308EA 51.4 EUR
215-3308 215-3152
Wash bottles, wide neck, DIY, Azlon®
Flessen Spuitflessen
LDPE with white one-piece PP closure
  • Indelibly printed in three languages (English, French and German)
  • Supplied with adhesive hazard labels
  • Good chemical and temperature resistance
Do-it-yourself wash bottles with blanc write-on panel.
Certificaten: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.