Spherical ground joints, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MS13UB FS13UB FS19UB FS29UB MS19UB MS29UB FS35UB MS35UB
201-8203EA 230 EUR
201-8203 201-8206 201-8207 201-8208 201-8204 201-8205 BIBBFS35UB BIBBMS35UB
Spherical ground joints, Quickfit®
Adapters Adapters met kogelgewrichten
Made of PYREX®, borosilicate glass.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm