Nessler cylinders
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CRC320 CRC322
BIBBCRC320EA 32.4 EUR
BIBBCRC320 BIBBCRC322
Nessler cylinders
Buisjes Nessler-buisjes
These Nessler cylinders are designed for colour comparison of water samples, foodstuffs, etc
  • Made of specially selected borosilicate glass tubing
  • Supplied in matched pairs for uniformity