Condensers, coil, reversible
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX3/35/SC CX3/23/SC
BIBBCX3/35/SCEA 328 EUR
BIBBCX3/35/SC BIBBCX3/23/SC
Condensers, coil, reversible
Condensors
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Highly versatile reflux condensers. Each end of the condenser is fitted with a different size cone thus reducing the need for adapters. With detachable plastic connectors for the attachment of flexible tubing.