Stopcocks, high vacuum
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SSV103 SSV104 SSV204
BIBBSSV103EA 163 EUR
BIBBSSV103 BIBBSSV104 201-8723
Stopcocks, high vacuum
Afsluitkranen
Borosilicate glass stopcocks specially designed for high vacuum applications.
  • All side-arms are 100 mm in length
  • Keys and barrels are precision ground together in matched pairs for optimum performance
  • Each key and barrel are numbered to ensure they are always paired together