Fluorescence cuvettes for flow-through meaurements
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
176751-85-40 131-10-40 176754-1015-40
HELL176751-85-40EA 1520 EUR
HELL176751-85-40 HELL131-10-40 HELL176754-1015-40
Fluorescence cuvettes for flow-through meaurements
Cuvetten Cuvetten voor spectrofotometers
The cuvettes are designed with a chamber to minimise bubble formation.
  • Screw connections tested to 5 bar
  • Cells with inlet/outlet tubes tested at 3 bar
  • Contact surfaces treated to reduce bubble adhesion within the chamber
Suitable for high precision analytical techniques.