QuEChERS extraction kits
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5982-7755 5982-5550 5982-5555 5982-5650 5982-5755 5982-5850 5982-6550 5982-6555 5982-6650 5982-6755 5982-7550 5982-7555 5982-7650 5982-5650CH 5982-5755CH 5982-5550CH 5982-5555CH 5982-1010
HEWL5982-7755EA 549 EUR
HEWL5982-7755 HEWL5982-5550 HEWL5982-5555 HEWL5982-5650 HEWL5982-5755 HEWL5982-5850 HEWL5982-6550 HEWL5982-6555 HEWL5982-6650 HEWL5982-6755 HEWL5982-7550 HEWL5982-7555 HEWL5982-7650 HEWL5982-5650CH HEWL5982-5755CH HEWL5982-5550CH HEWL5982-5555CH HEWL5982-1010
QuEChERS extraction kits
Kolommen voor chromatografie
Step 1: Extraction
Choose the extraction salt packet based on your method of analysis, AOAC or EN. The buffered extraction salts are amenable for more liable pesticides. Adding solvent and then salts to a comminuted fruit or vegetable sample (10 g or 15 g) enables you to extract the pesticides of interest into the organic layer. However, adding a food sample with a high percentage of water directly to the salts may create an exothermic reaction that can affect your analyte recoveries.
  • Available with or without 50 ml centrifuge tubes and caps
  • Include MgSO₄, NaCl, or other salts for