Reagent/centrifuge tubes, skirted base, with screw cap
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60.732.001 60.558.001 60.541.685 62.559.001 60.9922.243 60.549.001 60.732 62.543 60.540.686 60.549 60.551 60.551.001 60.558 60.9921.821 60.9921.829 60.9921.830 60.595.241 62.551.201 62.559
SARS60.732.001EA 0 EUR
SARS60.732.001 SARS60.558.001 SARS60.541.685 SARS62.559.001 SARS60.9922.243 SARS60.549.001 SARS60.732 SARS62.543 SARS60.540.686 SARS60.549 SARS60.551 SARS60.551.001 SARS60.558 SARS60.9921.821 SARS60.9921.829 SARS60.9921.830 SARS60.595.241 SARS62.551.201 SARS62.559
Reagent/centrifuge tubes, skirted base, with screw cap
Buisjes Centrifugebuisjes
PP, translucent oder PS, transparent with HDPE screw caps
  • Non sterile or sterile
  • Caps enclosed or assembled available
  • With or without graduation and writing space
Universal tubes for a wide range of applications.