HPLC columns, Pursuit™ UPS2,4
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A8120050X030H A8100030X020H A8100050X020H A8120030X020H A8120050X020H A8120100X020H A8120100X030H
VARIA8120050X030HEA 0 EUR
VARIA8120050X030H VARIA8100030X020H VARIA8100050X020H VARIA8120030X020H VARIA8120050X020H VARIA8120100X020H VARIA8120100X030H
HPLC columns, Pursuit™ UPS2,4
Kolommen voor chromatografie
Pursuit™ UPS 2,4 columns couple highly efficient, fast separations with superior ease-of-use. They are designed for solvent applications and samples where lifetime and back pressure are important concerns.
  • Minimal clogging and back pressure issues ensure easy of use and enhanced column life times
  • Ideal for use with 'greener' viscous solvents to reduce potential environmental hazards