Preparative HPLC columns, Dynamax
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R000083822 R000083824 R000083810 R000083821 R000083820 R000083840 R000083842 R000083814
VARIR000083822EA 0 EUR
VARIR000083822 VARIR000083824 VARIR000083810 VARIR000083821 VARIR000083820 VARIR000083840 VARIR000083842 VARIR000083814
Preparative HPLC columns, Dynamax
Kolommen voor chromatografie
The Dynamax preparative column hardware utilizes a patented dynamic axial compression (DAC) design and is the ideal format for the development and optimization of a high throughput or high yield purification.
  • Modular design with reusable end fittings reduces hardware costs
  • Three internal diameters of 10 mm, 21,4 mm and 41,4 mm for easy scale up
  • Integral guard column option for longer column lifetimes with complex samples
  • Kit 1 contains the end fittings for using the Dynamax column without a guard module
  • Kit 2 contains all the parts needed to operate with a
More Product Information
Order Now
Specificaties
Environmentally Preferable