Stirrer blades, PTFE, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST7/2 ST7/3
441-0359EA 55.3 EUR
441-0359 441-0360
Stirrer blades, PTFE, Quickfit®
Roerders Roerbladen
Stirrer blades made of PTFE, and they can be pivoted on the glass stirrer shaft to allow passage through small apertures.
  • Easily detachable for cleaning and replacement purposes
  • Resistance to chemical attack
Suitable for sweeping close to the vessel walls as they will not cause scratching.