Water estimators, heavy entrainers, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WE7/23/3 WE7/23/12
271-0019EA 208 EUR
271-0019 271-0018
Water estimators, heavy entrainers, Quickfit®
Onderdelen voor destillatie Destillatiesets
PYREX®, borosilicate glass
  • Graduated tube and Quickfit® precision joints
  • Also suitable for the estimation of volatile oil content
Estimator for the determination of the water content in petroleum products using a solvent entrainer wich is heavier than water.
BS 756
Certificaten: BS 756
Order Now
Specificaties
Environmentally Preferable