Rotoren voor microcentrifuge 5427 R
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5409702.009 5409704.001 5409706.004 5409708.007 5409710.001 5409712.004 5409714.007 5409715.003 5409700.006
521-1732EA 773 EUR
521-1732 521-1733 521-1734 521-1735 521-1736 521-1737 521-1738 521-1739 521-2066
Rotoren voor microcentrifuge 5427 R
Centrifuge Rotoren
Eppendorf rotors for centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight options available
Elevate your centrifugation experience with Eppendorf rotors crafted for centrifuge 5427 R. These precision rotors offer diverse capacities and speeds, providing flexibility for microcentrifugation or larger sample volumes. Accessories such as adaptors and buckets enhance customisation for specific applications. Maximise efficiency with reliable sample separation, meeting the demands of molecular biology, genomics, and more. Explore the full range of options to tailor your centrifuge setup, achieving reproducible results and optimising your laboratory workflow.