Culture flasks, narrow mouth, rimless
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1135/14D 1135/20D 1135/26D 1135/30D
391-0535EA 232 EUR
391-0535 391-0536 391-0537 391-0538
Culture flasks, narrow mouth, rimless
Kolven Cultuurflessen
Culture flask, made up of PYREX® borosilicate glass with white enamel markings. Compatible with shaker equipment.
  • Ideal for use in preparation of cultures
  • Long, rimless tube neck for use with Aluminium or PP closures
  • Excellent chemical and temperature resistance