Flasks acc. to Kjeldahl, with plain neck
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1200/12D 1200/16M 1200/04M 1200/10D
BIBB1200/12DEA 38.7 EUR
BIBB1200/12D BIBB1200/16M 511-0739 BIBB1200/10D
Flasks acc. to Kjeldahl, with plain neck
Kolven Kjeldahl-kolven
PYREX® borosilicate glass, plain neck.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes