Desiccators, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1590/02D 1591/02D 1593/02D 1594/02D 1612/03
BIBB1590/02DEA 245 EUR
BIBB1590/02D BIBB1591/02D BIBB1593/02D BIBB1594/02D BIBB1612/03
Desiccators, PYREX®
Exsiccatoren Exsiccatoren, glas
Ideal for vacuum and non vacuum applications.
  • Ground glass flanges
  • Borosilicate glass base and lid
  • Plates positioned on an internal ledge within the base