Solid phase extraction cartridges, Bond Elut™ Florisil
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12256022 12256030 12256038 12113049 12102024 12102050 12256014 12102109 12256046 12256047 12102129 12166014B 12162050B 12102159 52256014
VARI12256022EA 163 EUR
VARI12256022 551-0414 VARI12256038 VARI12113049 VARI12102024 551-0325 551-0408 VARI12102109 VARI12256046 VARI12256047 VARI12102129 VARI12166014B VARI12162050B VARI12102159 VARI52256014
Solid phase extraction cartridges, Bond Elut™ Florisil
Kolommen voor chromatografie
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from non-polar matrices.
  • Pesticide residue (PR) grade
  • Fast flow, ideal for viscous samples
The larger particle size of the sorbent enables flow for large sample volumes and is therefore an attractive alternative to silica if the sample matrix is particularly viscous.