Reagent/centrifuge tubes, flat base, with screw cap
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60.597.001 60.542.024 60.596.001 60.597 60.9922.113 60.9922.115 62.555 71.9923.610 60.542 60.542.007 60.596 60.9922.212 62.555.001
SARS60.597.001EA 0 EUR
SARS60.597.001 SARS60.542.024 SARS60.596.001 SARS60.597 SARS60.9922.113 SARS60.9922.115 SARS62.555 SARS71.9923.610 SARS60.542 SARS60.542.007 SARS60.596 SARS60.9922.212 SARS62.555.001
Reagent/centrifuge tubes, flat base, with screw cap
Buisjes Centrifugebuisjes
PP, translucent, PS or PC, transparent with PP screw caps
  • Non sterile or sterile
  • Caps enclosed or assembled available
  • With or without graduation
Universal tubes for a wide range of applications.