Spreaders, T-shaped
Leverancier: Sarstedt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SARS86.1569.005EA 72.5 EUR
SARS86.1569.005 SARS86.1569.001
Spreaders, T-shaped
Uitstrijkspatels Celspatels
Sterile inoculation spreaders made from blue PS. Ideal for spreading a bacterial suspension.