Reagent/centrifuge tubes, flat base, with push cap
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
58.537 58.487.100 58.536 58.485 58.490 58.631 62.486 58.485.001 58.489 58.491 58.505 62.470
SARS58.537EA 43.21 EUR
SARS58.537 SARS58.487.100 SARS58.536 SARS58.485 SARS58.490 SARS58.631 SARS62.486 SARS58.485.001 SARS58.489 SARS58.491 SARS58.505 SARS62.470
Reagent/centrifuge tubes, flat base, with push cap
Buisjes Centrifugebuisjes
PP, translucent, or PS, transparent with LDPE push caps
  • With or without push cap
  • Non sterile or sterile
  • With or without graduation
Universal tubes for a wide range of applications.