Adapters, receiver, plain bent, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RA9/22 RA9/33 RA9/44 RA9/11 RA9/23
271-0015EA 44.9 EUR
271-0015 271-0016 271-0017 271-0014 BIBBRA9/23
Adapters, receiver, plain bent, Quickfit®
Onderdelen voor destillatie Destillatieadapters
Receiver, delivery, PYREX® borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Plain bend without vent.